Abnova
Manufacturer Code:MAB12035
Catalog # 89-110-179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SPM348 |
Immunogen | Recombinant GST fusion protein corresponding to mouse Cdkn1b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of Cdkn1b.
For Research Use Only
Protein Aliases: AA408329; AI843786; Cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor p27; Kip1; p27; p27Kip1
Gene Aliases: AA408329; AI843786; Cdkn1b; Kip1; p27; p27Kip1
UniProt ID: (Mouse) P46414
Entrez Gene ID: (Mouse) 12576
Molecular Function: enzyme modulator kinase inhibitor kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.