Novus Biologicals
Manufacturer Code:NBP303448100UL
Catalog # NB092427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-326 of human Cdk6 (NP_001250.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell division protein kinase 6; Cell division protein kinase 6 cyclin-dependent kinase 6 EC 2.7.11 EC 2.7.11.22 PLSTIREMGC59692 Serine/threonine-protein kinase PLSTIRE; Cyclin-dependent kinase 6; Serine/threonine-protein kinase PLSTIRE
Gene Aliases: CDK6; CDKN6; MCPH12; PLSTIRE
UniProt ID: (Human) Q00534
Entrez Gene ID: (Human) 1021
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.