Invitrogen
Manufacturer Code:LFPA0132
Catalog # APL0132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | His-tagged recombit human CDK5 protein purified from baculovirus expressed insect cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell division protein kinase 5; Cell division protein kinase 5 Crk6 Cyclin dependent kinase 5; CR6 protein kinase; CRK6; cyclin-dependent kinase 5; Cyclin-dependent-like kinase 5; protein kinase CDK5 splicing; Serine/threonine-protein kinase PSSALRE; Tau protein kinase II catalytic subunit; TPKII catalytic subunit
Gene Aliases: AW048668; CDK5; CDKN5; Crk6; LIS7; PSSALRE
UniProt ID: (Human) Q00535, (Mouse) P49615
Entrez Gene ID: (Human) 1020, (Mouse) 12568
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.