Invitrogen
Manufacturer Code:MA511472
Catalog # PIMA511472
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A17.1.1 |
Immunogen | C-Terminal 2/3 of Xenopus CDC2 expressed in E. coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CDK1; CDK1-A; cell cycle controller CDC2; cell cycle controller CDC2 cell division control protein 2 homolog cell division cycle 2 G1 to S and G2 to M cell division protein kinase 1 p34 protein kinase; cell cycle p34 CDC2 kinase protein; Cell division control protein 2 homolog; Cell division control protein 2 homolog 1; Cell division control protein 2-A; cell division cycle 2 homolog A; cell division cycle 2 like; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; Cell division protein kinase 1; Cell division protein kinase 1-A; Cyclin-dependent kinase 1; Cyclin-dependent kinase 1-A; maturation-promoting factor; p34 protein kinase; p34 protein kinase 1; unnamed protein product
Gene Aliases: cdc-2; CDC2; cdc2-a; CDC28A; Cdc2a; cdc2x1.1; CDK1; cdk1-a; cdk1-b; CDKN1; MPF; p34<CDC2>; P34CDC2; PSTAIR; xcdc2; XELAEV_18036802mg
UniProt ID: (Chicken) P13863, (Human) P06493, (Rat) P39951, (Mouse) P11440, (Xenopus) P35567
Entrez Gene ID: (Chicken) 396252, (Human) 983, (Guinea pig) 100735946, (Rat) 54237, (Mouse) 12534, (Xenopus) 380246
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.