Novus Biologicals
Manufacturer Code:NBP246192
Catalog # NBP246192
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A4 |
Immunogen | Full length human recombinant protein of human CDC6(NP_001245) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cdc18-related protein; CDC18LHsCDC18 Cdc18-related protein CDC6 (cell division cycle 6 S. cerevisiae) homolog CDC6 cell division cycle 6 homolog CDC6 cell division cycle 6 homolog (S. cerevisiae) CDC6-related protein cell division control protein 6 homolog cell division cycle 6 homolog (S. cerevisiae) cell division cycle 6 protein HsCdc18 HsCDC6 p62(cdc6); CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6 homolog; HsCdc18; HsCDC6; p62(cdc6)
Gene Aliases: CDC18L; CDC6; HsCDC18; HsCDC6
UniProt ID: (Human) Q99741
Entrez Gene ID: (Human) 990
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.