Invitrogen
Manufacturer Code:PA577901
Catalog # PIPA577901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human CDC6. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC18(S.pombe); Cdc18-related protein; CDC18L HsCDC18 HsCDC6 CDC6 p62(cdc6) cdc18-related protein CDC6-related protein CDC6 cell division cycle 6 homolog cell division control protein 6 homolog cell division cycle 6 homolog (S. cerevisiae); CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6 homolog; HsCdc18; HsCDC6; p62(cdc6)
Gene Aliases: CDC18L; CDC6; HsCDC18; HsCDC6
UniProt ID: (Human) Q99741, (Mouse) O89033
Entrez Gene ID: (Human) 990, (Mouse) 23834, (Rat) 360621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.