Invitrogen
Manufacturer Code:MA124748
Catalog # PIMA124748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DCS-193 |
Immunogen | Recombit full length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cyclin dependent kinase 25c; Dual specificity phosphatase Cdc25C; M-phase inducer phosphatase 3; mitosis inducer CDC25; phosphotyrosine phosphatase; protein phosphatase 1, regulatory subunit 60
Gene Aliases: CDC25; CDC25C; PPP1R60
UniProt ID: (Human) P30307
Entrez Gene ID: (Human) 995
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.