Invitrogen
Manufacturer Code:PA585509
Catalog # PIPA585509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human CDC25C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC25 antibody CDC25C antibody mitosis inducer CDC25 antibody dual specificity phosphatase CDC25C antibody M-phase inducer phosphatase 3 antibody cell division cycle 25C antibody protein phosphatase 1 regulatory subunit 60 antibody phosphotyrosine phosphatase antibody cell division cycle 25 homolog C (S. pombe) antibody; Dual specificity phosphatase Cdc25C; M-phase inducer phosphatase 3; mitosis inducer CDC25; phosphotyrosine phosphatase; protein phosphatase 1, regulatory subunit 60
Gene Aliases: CDC25; CDC25C; Cdc25m1; PPP1R60
UniProt ID: (Human) P30307, (Mouse) Q99KG6
Entrez Gene ID: (Human) 995, (Mouse) 12532, (Rat) 307511
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.