Abnova
Manufacturer Code:MAB3238
Catalog # 89-111-286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | GL1 |
Immunogen | Native Cd86 from LPS-activated mouse B cells. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rat monoclonal antibody raised against Cd86.
For Research Use Only
Protein Aliases: Activation B7-2 antigen; B7; B7-2; B7.2; B70; Cd28l2; CD86; CLS1; early T cell costimulatory molecule-1; Early T-cell costimulatory molecule 1; ETC-1; Ly-58; Ly58; MB7; MB7-2; T-lymphocyte activation antigen CD86; TS/A-2
Gene Aliases: B7; B7-2; B7.2; B70; Cd28l2; Cd86; CLS1; ETC-1; Ly-58; Ly58; MB7; MB7-2; TS/A-2
UniProt ID: (Mouse) P42082
Entrez Gene ID: (Mouse) 12524
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.