Abnova
Manufacturer Code:MAB5791
Catalog # 89-112-462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Chinese hamster |
Class | Monoclonal |
Type | Antibody |
Clone | 2F7 |
Immunogen | Native purified Cd81 from mouse epithelial cell line PAM212. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Do not freeze.<br />Avoid prolonged exposure to light.<br />Aliquot to avoid repeated freezing and thawing. |
Hamster monoclonal antibody raised against native Cd81.
For Research Use Only
Protein Aliases: 26 kDa cell surface protein TAPA-1; CD81; CD81 antigen; Tapa-1; Tapa1; Target of the antiproliferative antibody 1; Tspan28
Gene Aliases: Cd81; Tapa-1; Tapa1; Tspan28
UniProt ID: (Mouse) P35762
Entrez Gene ID: (Mouse) 12520
Molecular Function: cell adhesion molecule membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.