Abnova
Manufacturer Code:MAB16361
Catalog # 89-999-161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | IM7 |
Immunogen | Mouse Cd44. |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AU023126; AW121933; AW146109; CD44; CD44 antigen; ECMR-III; Extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; HERMES; Hermes antigen; HUTCH-I; Hyaluronate receptor; Ly-24; Lymphocyte antigen 24; PGP-1; PGP-I; Phagocytic glycoprotein 1; Phagocytic glycoprotein I
Gene Aliases: AU023126; AW121933; AW146109; Cd44; HERMES; Ly-24; Pgp-1
UniProt ID: (Mouse) Q62061
Entrez Gene ID: (Mouse) 12505
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.