Abnova
Manufacturer Code:PAB9097
Catalog # 89-134-151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acid 1-70 of rat Ccl5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Ccl5.
For Research Use Only
Protein Aliases: C-C motif chemokine 5; Rantes; regulated upon activation normal T-cell expressed and secreted; Scya5; SIS-delta; small inducible cytokine A5; small-inducible cytokine A5; T-cell-specific protein RANTES
Gene Aliases: Rantes; Scya5
Entrez Gene ID: (Rat) 81780
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.