Abnova
Manufacturer Code:PAB19112
Catalog # 89-120-644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Ccl4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 200&muL of deionized water and concentration will be 500&mu/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ccl4.
For Research Use Only
Protein Aliases: ACT-2; ACT2; AT744.1; C-C motif chemokine 4; Immune activation protein 2; Macrophage inflammatory protein 1-beta; MIP-1 beta; MIP-1-beta; MIP-1B; Mip1b; Protein H400; Scya4; SIS-gamma; small inducible cytokine A4; Small-inducible cytokine A4
Gene Aliases: Act-2; AT744.1; Ccl4; MIP-1B; Mip1b; Scya4
UniProt ID: (Mouse) P14097
Entrez Gene ID: (Mouse) 20303
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.