Abnova
Manufacturer Code:PAB15518
Catalog # 89-117-629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 30-80 of rat Ccl2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ccl2.
For Research Use Only
Protein Aliases: C-C motif chemokine 2; immediate-early serum-responsive JE protein; Immediate-early serum-responsive protein JE; MCP-1; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; Monocyte chemotactic protein 1; Scya2; Sigje; small inducible cytokine A2; small inducible gene JE; Small-inducible cytokine A2
Gene Aliases: Ccl2; Je; MCP-1; Mcp1; Scya2; Sigje
UniProt ID: (Rat) P14844
Entrez Gene ID: (Rat) 24770
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.