R&D Systems
Manufacturer Code:MAB6706
Catalog # MAB6706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 682708 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Caveolin-3 Met1-Pro83 Accession # P56539 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAV3 caveolin 3 caveolin-3 Caveolin3 LGMD1C LGMD1CMGC126100 LQT9 LQT9MGC126101 M-caveolin MGC126129 VIP21 VIP-21; Caveolin-3; M-caveolin
Gene Aliases: CAV3; LGMD1C; LQT9; VIP-21; VIP21
UniProt ID: (Human) P56539
Entrez Gene ID: (Human) 859
Molecular Function:
G-protein modulator
enzyme modulator
membrane traffic protein
structural protein
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.