Invitrogen
Manufacturer Code:PA519116
Catalog # PIPA519116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EHTDLEAQIVKDIH) corresponding to the N-terminus amino acids of CAV3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CAV3 caveolin 3 LGMD1C LQT9 MGC126100 MGC126101 MGC126129 VIP-21 VIP21 M-caveolin; Caveolin-3; M-caveolin; structural protein
Gene Aliases: AI385751; Cav-3; CAV3; LGMD1C; LQT9; M-cav; VIP-21; VIP21
UniProt ID: (Pig) Q3ZDQ5, (Human) P56539, (Rat) P51638, (Mouse) P51637
Entrez Gene ID: (Pig) 641358, (Human) 859, (Rat) 29161, (Mouse) 12391
Molecular Function:
G-protein modulator
enzyme modulator
membrane traffic protein
structural protein
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.