Novus Biologicals
Manufacturer Code:NBP128917
Catalog # NBP128917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence EHTDLEAQIVKDIH-C corresponding to N-Terminus according to NP_203123.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: caveolin 3 caveolin-3 LGMD1CMGC126100 LQT9MGC126101 M-caveolin MGC126129 VIP21 VIP-21; Caveolin-3; M-caveolin
Gene Aliases: CAV3; LGMD1C; LQT9; VIP-21; VIP21
UniProt ID: (Human) P56539
Entrez Gene ID: (Human) 859
Molecular Function:
G-protein modulator
enzyme modulator
membrane traffic protein
structural protein
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.