Invitrogen
Manufacturer Code:MA535600
Catalog # PIMA535600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0323 |
Immunogen | A synthesized peptide derived from human CAV2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI447843 CAV Cav2 Caveolin-2 MGC12294 Phospho-CAV2; caveolae protein, 20-kD; caveolin 2 isoform a and b; Caveolin-2
Gene Aliases: AI447843; CAV; CAV2
UniProt ID: (Human) P51636, (Mouse) Q9WVC3
Entrez Gene ID: (Human) 858, (Mouse) 12390
Molecular Function:
G-protein modulator
enzyme modulator
membrane traffic protein
structural protein
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.