Novus Biologicals
Manufacturer Code:NBP229935
Catalog # NBP229935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This CAV2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 11-44 amino acids from the N-terminal region of human CAV2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAV caveolae protein 20-kD caveolin 2 caveolin 2 isoform a and b caveolin-2 MGC12294; caveolae protein, 20-kD; caveolin 2 isoform a and b; Caveolin-2
Gene Aliases: CAV; CAV2
UniProt ID: (Human) P51636
Entrez Gene ID: (Human) 858
Molecular Function: G-protein modulator enzyme modulator membrane traffic protein structural protein transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.