R&D Systems
Manufacturer Code:BAF5788
Catalog # BAF5788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Caveolin-2 Met1-Lys86 Accession # P51636 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
Protein Aliases: CAV CAV2 caveolae protein 20-kD caveolin 2 caveolin 2 isoform a and b caveolin-2 Caveolin2 MGC12294; caveolae protein, 20-kD; caveolin 2 isoform a and b; Caveolin-2
Gene Aliases: CAV; CAV2
UniProt ID: (Human) P51636
Entrez Gene ID: (Human) 858
Molecular Function: G-protein modulator enzyme modulator membrane traffic protein structural protein transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.