Novus Biologicals
Manufacturer Code:NB100615
Catalog # NB100615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7C8 |
Immunogen | Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAV caveolae protein 22kD caveolin 1 caveolae protein 22kDa cell growth-inhibiting protein 32 MSTP085; caveolin 1, caveolae protein, 22kDa; Caveolin-1; cell growth-inhibiting protein 32
Gene Aliases: BSCL3; CAV; CAV1; CGL3; LCCNS; MSTP085; PPH3; VIP21
UniProt ID: (Human) Q03135
Entrez Gene ID: (Human) 857
Molecular Function:
G-protein modulator
enzyme modulator
membrane traffic protein
structural protein
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.