Invitrogen
Manufacturer Code:PA517447
Catalog # PIPA517447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Glu20 of human caveolin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAV1; caveolin 1, caveolae protein, 22kDa; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; Caveolin-1; cell growth-inhibiting protein 32; OTTHUMP00000196475 caveolin-1 cell growth-inhibiting protein 32; variant a; variant b
Gene Aliases: BSCL3; CAV; Cav-1; CAV1; Caveolin-1; CGL3; LCCNS; MSTP085; PPH3; VIP21; wu:fc07c04
UniProt ID: (Pig) Q6RVA9, (Chicken) P35431, (Bovine) P79132, (Human) Q03135, (Mouse) P49817
Entrez Gene ID: (Pig) 404693, (Chicken) 373996, (Bovine) 281040, (Human) 857, (Mouse) 12389, (Rat) 25404, (Chinese hamster) 100753021, (Zebrafish) 323695
Molecular Function:
G-protein modulator
enzyme modulator
membrane traffic protein
structural protein
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.