Novus Biologicals
Manufacturer Code:NBP122444UV
Catalog # NBP122444UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S55-10 |
Immunogen | Fusion protein amino acids 1019-1293 (II-III loop) of human Cav3.2 |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CACNA1HB calcium channel voltage-dependent T type alpha 1H subunit calcium channel voltage-dependent T type alpha 1Hb subunit Cav3.2 ECA6 EIG6 FLJ90484 Low-voltage-activated calcium channel alpha1 3.2 subunit low-voltage-activated calcium channel alpha13.2 subunit voltage dependent t-type calcium channel alpha-1H subunit voltage-dependent T-type calcium channel subunit alpha-1H voltage-gated calcium channel alpha subunit Cav3.2 voltage-gated calcium channel alpha subunit CavT.2 Voltage-gated calcium channel subunit alpha Cav3.2; calcium channel, voltage-dependent, T type, alpha 1H subunit; calcium channel, voltage-dependent, T type, alpha 1Hb subunit; Low-voltage-activated calcium channel alpha1 3.2 subunit; low-voltage-activated calcium channel alpha13.2 subunit; voltage dependent t-type calcium channel alpha-1H subunit; Voltage-dependent T-type calcium channel subunit alpha-1H; voltage-gated calcium channel alpha subunit Cav3.2; voltage-gated calcium channel alpha subunit CavT.2; Voltage-gated calcium channel subunit alpha Cav3.2
Gene Aliases: CACNA1H; CACNA1HB; Cav3.2; ECA6; EIG6
UniProt ID: (Human) O95180
Entrez Gene ID: (Human) 8912
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.