Novus Biologicals
Manufacturer Code:NBP212894
Catalog # NBP212894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S38-8 |
Immunogen | Fusion protein amino acids 2025-2161 of rat Cav1.3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CACN4calcium channel neuroendocrine/brain-type alpha 1 subunit CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D Calcium channel L type alpha-1 polypeptide isoform 2 calcium channel voltage-dependent L type alpha 1D subunit Cav1.3 CCHL1A2voltage-gated calcium channel alpha 1 subunit L type alpha-1 polypeptide voltage-gated calcium channel alpha subunit Cav1.3 Voltage-gated calcium channel subunit alpha Cav1.3; Calcium channel, L type, alpha-1 polypeptide, isoform 2; calcium channel, neuroendocrine/brain-type, alpha 1 subunit; calcium channel, voltage-dependent, L type, alpha 1D subunit; Voltage-dependent L-type calcium channel subunit alpha-1D; voltage-gated calcium channel alpha 1 subunit; voltage-gated calcium channel alpha subunit Cav1.3; Voltage-gated calcium channel subunit alpha Cav1.3
Gene Aliases: CACH3; CACN4; CACNA1D; CACNL1A2; Cav1.3; CCHL1A2; PASNA; SANDD
UniProt ID: (Human) Q01668
Entrez Gene ID: (Human) 776
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.