Novus Biologicals
Manufacturer Code:NBP122439AF647
Catalog # NBP122439Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S57-46 |
Immunogen | Fusion protein amino acids 1507-1733 (intracellular carboxyl terminus) of rabbit Cav1.2 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha-1 subunit calcium channel voltage-dependent L type alpha 1C subunit cardic dihydropyridine-sensitive alpha-1 subunit L type alpha 1 polypeptide isoform 1 cardic muscle L type alpha-1 polypeptide isoform 1 cardiac muscle splice variant 10*; calcium channel, cardic dihydropyridine-sensitive, alpha-1 subunit; Calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle; calcium channel, voltage-dependent, L type, alpha 1C subunit; DHPR, alpha-1 subunit; voltage-dependent L-type Ca2+ channel alpha 1 subunit; Voltage-dependent L-type calcium channel subunit alpha-1C; Voltage-gated calcium channel subunit alpha Cav1.2; voltage-gated L-type calcium channel Cav1.2 alpha 1 subunit, splice variant 10*
Gene Aliases: CACH2; CACN2; CACNA1C; CACNL1A1; CaV1.2; CCHL1A1; LQT8; TS
UniProt ID: (Human) Q13936
Entrez Gene ID: (Human) 775
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.