Invitrogen
Manufacturer Code:PA518991
Catalog # PIPA518991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RARGRPSEEELQD) corresponding to the C-terminus amino acids of CACNA1C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CACNA1C calcium channel voltage-dependent L type alpha 1C subunit CACH2 CACN2 CACNL1A1 CCHL1A1 CaV1.2 MGC120730 TS DHPR alpha-1 subunit calcium channel L type alpha 1 polypeptide isoform 1 cardic muscle calcium channel cardic dihydropyridine-sensitive alpha-1 subunit voltage-dependent L-type calcium channel alpha 1C subunit voltage-gated L-type calcium channel Cav1.2 alpha 1 subunit splice variant 10* voltage-gated calcium channel alpha subunit Cav1.2 LTCC; calcium channel, cardic dihydropyridine-sensitive, alpha-1 subunit; Calcium channel, L type, alpha-1 polypeptide, isoform 1, cardiac muscle; calcium channel, voltage-dependent, L type, alpha 1C subunit; DHPR, alpha-1 subunit; voltage-dependent L-type Ca2+ channel alpha 1 subunit; Voltage-dependent L-type calcium channel subunit alpha-1C; Voltage-gated calcium channel subunit alpha Cav1.2; voltage-gated L-type calcium channel Cav1.2 alpha 1 subunit, splice variant 10*
Gene Aliases: CACH2; CACN2; CACNA1C; CACNL1A1; CaV1.2; CCHL1A1; LQT8; TS
UniProt ID: (Human) Q13936
Entrez Gene ID: (Human) 775
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.