Novus Biologicals
Manufacturer Code:NBP156308
Catalog # NBP156308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CAD(carbamoyl-phosphate synthetase 2 aspartate transcarbamylase and dihydroorotase) The peptide sequence was selected from the middle region of CAD. Peptide sequence SVSNNNRTSPSKPPYFDWMKKPAYPAQPQPGKTRT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cad-PA; cad-PB; cad-PC; CAUDAL; CG1759 gene product from transcript CG1759-RA 38E.19 anon-WO2004063362.83 Cad caudal cd CG1759 DmelCG1759 S67; CG1759-PA; CG1759-PB; CG1759-PC; Homeotic protein caudal; LD29596p
Gene Aliases: 38E.19; anon-WO2004063362.83; CAD; cd; CG1759; Dmel\CG1759; Dmel_CG1759; S67
UniProt ID: (Fruit fly) P09085
Entrez Gene ID: (Fruit fly) 35341
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.