Novus Biologicals
Manufacturer Code:NBP212085R
Catalog # NBP212085R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0133-9G24 |
Immunogen | Human Cathepsin V |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cathepsin L2; cathepsin L2 Cathepsin U Cathepsin V CATL2 CTSUcathepsin L2 preproprotein CTSVMGC125957 EC 3.4.22 EC 3.4.22.43; cathepsin L2, preproprotein; Cathepsin U; Cathepsin V
Gene Aliases: CATL2; CTSL2; CTSU; CTSV; UNQ268/PRO305
UniProt ID: (Human) O60911
Entrez Gene ID: (Human) 1515
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.