R&D Systems
Manufacturer Code:AF1080SP
Catalog # AF1080SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Cathepsin V Val18-Val334 Accession # O60911 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cathepsin L2; cathepsin L2 Cathepsin U Cathepsin V CATL2 CTSL2 CTSUcathepsin L2 preproprotein CTSVMGC125957 EC 3.4.22 EC 3.4.22.43; cathepsin L2, preproprotein; Cathepsin U; Cathepsin V
Gene Aliases: CATL2; CTSL2; CTSU; CTSV; UNQ268/PRO305
UniProt ID: (Human) O60911
Entrez Gene ID: (Human) 1515
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.