Proteintech
Manufacturer Code:184421AP150UL
Catalog # 50-173-2154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cathepsin V Fusion Protein Ag13284 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cathepsin L2; cathepsin L2 Cathepsin U Cathepsin V CATL2 CTSL2 CTSU CTSV; cathepsin L2, preproprotein; Cathepsin U; Cathepsin V
Gene Aliases: CATL2; CTSL2; CTSU; CTSV; UNQ268/PRO305
UniProt ID: (Human) O60911
Entrez Gene ID: (Human) 1515
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.