Invitrogen
Manufacturer Code:PA581368
Catalog # PIPA581368
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse Cathepsin L/CTSL protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1190035F06Rik Ctsl1 MEP fs nkt; Cat L; Cathepsin L; Cathepsin L heavy chain; Cathepsin L light chain; Cathepsin L1; furless; Major excreted protein; MEP; nackt; p39 cysteine proteinase; Procathepsin L
Gene Aliases: 1190035F06Rik; Ctsl; Ctsl1; fs; MEP; nkt
UniProt ID: (Mouse) P06797
Entrez Gene ID: (Mouse) 13039
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.