Proteintech
Manufacturer Code:112391AP150UL
Catalog # 50-172-6994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cathepsin K Fusion Protein Ag1761 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cat K; Cathepsin K; Cathepsin K Cathepsin O Cathepsin O2 Cathepsin X CTS02 CTSK CTSO CTSO1 CTSO2 PKND PYCD; Cathepsin O; cathepsin O1; Cathepsin O2; Cathepsin X; minisatellite 10q detected by probe MMS10
Gene Aliases: AI323530; catK; CTS02; CTSK; CTSO; CTSO1; CTSO2; MMS10-Q; Ms10q; PKND; PYCD
UniProt ID: (Human) P43235, (Rat) O35186, (Mouse) P55097
Entrez Gene ID: (Human) 1513, (Rat) 29175, (Mouse) 13038
Molecular Function:
cysteine protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.