Invitrogen
Manufacturer Code:MA535614
Catalog # PIMA535614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1276 |
Immunogen | A synthesized peptide derived from human Cathepsin L/V/K/H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ACC4 ACC-4 ACC5 ACC-5 AI323530 AL022844 Cathepsin B3 Cathepsin BA Cathepsin H Cathepsin H heavy chain Cathepsin H light chain Cathepsin H mini chain Cathepsin K Cathepsin L2 Cathepsin O Cathepsin O2 Cathepsin U Cathepsin V Cathepsin X catK CATL2 CPSB CTS02 Ctsh Ctsk CTSL2 CTSO CTSO1 CTSO2 CTSU CTSV MMS10-Q Ms10q PKND Pro-cathepsin H PYCD UNQ268/PRO305; Cat H; Cat K; Cathepsin B3; Cathepsin BA; Cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; Cathepsin K; minisatellite 10q detected by probe MMS10; Pro-cathepsin H
Gene Aliases: AI323530; AL022844; catK; Ctsh; Ctsk; MMS10-Q; Ms10q
UniProt ID: (Rat) P00786, (Mouse) P49935, (Rat) O35186, (Mouse) P55097
Entrez Gene ID: (Rat) 25425, (Mouse) 13036, (Rat) 29175, (Mouse) 13038
Molecular Function:
cysteine protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.