Novus Biologicals
Manufacturer Code:NBP212080
Catalog # NBP212080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0165-8J11 |
Immunogen | Mouse Cathepsin H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ACC-4 ACC-5 aleurain cathepsin B3 cathepsin BA cathepsin H CPSB DKFZp686B24257 EC 3.4.22 EC 3.4.22.16 MGC1519 minichain N-benzoylarginine-beta-naphthylamide hydrolase; aleurain; cathepsin B3; cathepsin BA; Cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; N-benzoylarginine-beta-naphthylamide hydrolase; Pro-cathepsin H
Gene Aliases: ACC-4; ACC-5; ACC4; ACC5; CPSB; CTSH
UniProt ID: (Human) P09668
Entrez Gene ID: (Human) 1512
Molecular Function:
cysteine protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.