Invitrogen
Manufacturer Code:PA575439
Catalog # PIPA575439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide to amino acids 171-210 of Human Cathepsin H. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aleurain; Cat H; Cathepsin B3; Cathepsin BA; Cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; N-benzoylarginine-beta-naphthylamide hydrolase; Pro-cathepsin H; Pro-cathepsin H Cathepsin H Cathepsin H heavy chain CTSH CPSB CATH
Gene Aliases: ACC-4; ACC-5; ACC4; ACC5; AL022844; CPSB; CTSH
UniProt ID: (Human) P09668, (Rat) P00786, (Mouse) P49935
Entrez Gene ID: (Human) 1512, (Rat) 25425, (Mouse) 13036
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.