Invitrogen
Manufacturer Code:PA547041
Catalog # PIPA547041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Cathepsin H Glu22-Val333 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cat H; Cathepsin B3; Cathepsin BA; Cathepsin H; Cathepsin H Cathepsin H mini chain Cathepsin H heavy chain Cathepsin H light chain CPSB CTSH; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; Pro-cathepsin H
Gene Aliases: AL022844; Ctsh
UniProt ID: (Mouse) P49935
Entrez Gene ID: (Mouse) 13036
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.