R&D Systems
Manufacturer Code:AF1013
Catalog # AF1013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin H Glu22-Val333 Accession # Q3UCD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACC-4 ACC-5 aleurain cathepsin B3 cathepsin BA cathepsin H CPSB CTSH DKFZp686B24257 EC 3.4.22 EC 3.4.22.16 MGC1519 minichain N-benzoylarginine-beta-naphthylamide hydrolase; aleurain; cathepsin B3; cathepsin BA; Cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; N-benzoylarginine-beta-naphthylamide hydrolase; Pro-cathepsin H
Gene Aliases: ACC-4; ACC-5; ACC4; ACC5; CPSB; CTSH
UniProt ID: (Human) P09668
Entrez Gene ID: (Human) 1512
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.