Novus Biologicals
Manufacturer Code:NBP29011950UL
Catalog # NB021702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 10 |
Immunogen | Recombinant Human Cathepsin D protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Cathepsin D; cathepsin D cathepsin D (lysosomal aspartyl protease) CPSD EC 3.4.23 EC 3.4.23.5 lysosomal aspartyl peptidase lysosomal aspartyl protease MGC2311 neuronal 10; Cathepsin D heavy chain; Cathepsin D light chain; ceroid-lipofuscinosis, neuronal 10; epididymis secretory sperm binding protein Li 130P; lysosomal aspartyl peptidase; lysosomal aspartyl protease
Gene Aliases: CLN10; CPSD; CTSD; HEL-S-130P
UniProt ID: (Human) P07339
Entrez Gene ID: (Human) 1509
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.