R&D Systems
Manufacturer Code:AF1034SP
Catalog # AF1034SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin C/DPPI Asp25-Leu462 Accession # P97821 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cathepsin C; cathepsin CEC 3.4.14.1 Cathepsin J CPPIHMS CTSC dipeptidyl peptidase 1 Dipeptidyl peptidase I Dipeptidyl transferase dipeptidyl-peptidase I DPP1 DPPI DPP-I JP JPD PALS PLS; Cathepsin J; Dipeptidyl peptidase 1; Dipeptidyl peptidase 1 exclusion domain chain; Dipeptidyl peptidase 1 heavy chain; Dipeptidyl peptidase 1 light chain; Dipeptidyl peptidase I; Dipeptidyl peptidase I exclusion domain chain; Dipeptidyl peptidase I heavy chain; Dipeptidyl peptidase I light chain; Dipeptidyl transferase; dipeptidyl-peptidase I; DPP-I
Gene Aliases: CPPI; CTSC; DPP-I; DPP1; DPPI; HMS; JP; JPD; PALS; PDON1; PLS
UniProt ID: (Human) P53634
Entrez Gene ID: (Human) 1075
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.