Novus Biologicals
Manufacturer Code:NBP290401AF532
Catalog # NB073096
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse Cathepsin B (P10605 Met1-Phe339). |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: amyloid precursor protein secretase; APP secretase; APP secretase APPS cathepsin B Cathepsin B1 CPSBamyloid precursor protein secretase cysteine protease EC 3.4.22 EC 3.4.22.1; APPS; Cathepsin B; Cathepsin B heavy chain; Cathepsin B light chain; Cathepsin B1; cysteine protease
Gene Aliases: APPS; CPSB; CTSB
UniProt ID: (Human) P07858
Entrez Gene ID: (Human) 1508
Molecular Function:
cysteine protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.