Invitrogen
Manufacturer Code:PA576029
Catalog # PIPA576029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human Cathelicidin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAP-18; Cathelicidin antimicrobial peptide; Cathelicidin antimicrobial peptide 18 kDa cationic antimicrobial protein CAP-18 hCAP-18 CAP18 FALL39 HSD26; Cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cramp; FALL-39 peptide antibiotic
Gene Aliases: CAMP; CAP-18; CAP18; Cnlp; CRAMP; FALL-39; FALL39; HSD26; LL37; MCLP
UniProt ID: (Human) P49913, (Mouse) P51437
Entrez Gene ID: (Human) 820, (Mouse) 12796
Molecular Function: antibacterial response protein defense/immunity protein enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.