Invitrogen
Manufacturer Code:PA5110987
Catalog # PIPA5110987
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human Cathelicidin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18 kDa cationic antimicrobial protein; 18 kDa cationic antimicrobial protein Antibacterial peptide FALL-39 Antibacterial peptide LL-37 Camp CAP18 CAP-18 Cathelicidin antimicrobial peptide Cathelin-like protein Cathelin-related antimicrobial peptide CLP Cnlp Cramp FALL39 FALL-39 FALL-39 peptide antibiotic hCAP18 hCAP-18 HSD26 LL37 MCLP; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAP-18; Cathelicidin antimicrobial peptide; FALL-39 peptide antibiotic
Gene Aliases: CAMP; CAP-18; CAP18; CRAMP; FALL-39; FALL39; HSD26; LL37
UniProt ID: (Human) P49913
Entrez Gene ID: (Human) 820
Molecular Function:
antibacterial response protein
defense/immunity protein
enzyme modulator
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.