Novus Biologicals
Manufacturer Code:NBP29275320UL
Catalog # NB027376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 150-300 of human CTNNA2 (NP_004380.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha N-catenin; Alpha-catenin-related protein; Alpha-catenin-related protein cancer/testis antigen 114 CAPRrelated catenin (cadherin-associated protein) alpha 2 catenin alpha-2 CTNR DKFZp686H02198; alpha-N-catenin; cadherin-associated protein, related; cancer/testis antigen 114; catenin (cadherin-associated protein), alpha 2; Catenin alpha-2
Gene Aliases: CAP-R; CAPR; CT114; CTNNA2; CTNR
UniProt ID: (Human) B7Z2W7
Entrez Gene ID: (Human) 1496
Molecular Function:
actin family cytoskeletal protein
cell adhesion molecule
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.