Novus Biologicals
Manufacturer Code:NBP298329B
Catalog # NB081985
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Caspr2 (NP_001004357.2 Met 1-Ser 1262). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AUTS15 CASPR2DKFZp781D1846 CDFE Cell recognition molecule Caspr2 contactin associated protein-like 2 contactin-associated protein 2 contactin-associated protein-like 2 homolog of Drosophila neurexin IV KIAA0868 NRXN4Caspr2 PTHSL1; Cell recognition molecule Caspr2; Contactin-associated protein-like 2; homolog of Drosophila neurexin IV
Gene Aliases: AUTS15; CASPR2; CDFE; CNTNAP2; KIAA0868; NRXN4; PTHSL1
UniProt ID: (Human) Q9UHC6
Entrez Gene ID: (Human) 26047
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.