Proteintech
Manufacturer Code:103801AP150UL
Catalog # 50-172-6260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Caspase 9/p35/p10 Fusion Protein Ag0404 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 25 kDa caspase-9 dominant negative protein; APAF 3 APAF3 Apoptotic protease Mch 6 CASP 9 CASP9 Caspase 9 CASPASE 9c Caspase9 Caspase-9 ICE LAP6 ICE like apoptotic protease 6 MCH6; APAF-3; apoptotic protease activating factor 3; Apoptotic protease Mch-6; Apoptotic protease-activating factor 3; CASP-9; caspase 9, apoptosis-related cysteine peptidase; Caspase-9; caspase-9 dominant negative form; Caspase-9 subunit p10; Caspase-9 subunit p35; caspase-9-carboxyl-terminal divergent; ICE-LAP6; ICE-like apoptotic protease 6; protein phosphatase 1, regulatory subunit 56
Gene Aliases: AI115399; APAF-3; APAF3; AW493809; CASP-9; Casp-9-CTD; CASP9; Casp9_v1; Caspase-9; ICE-LAP6; MCH6; PPP1R56
UniProt ID: (Human) P55211, (Mouse) Q8C3Q9
Entrez Gene ID: (Human) 842, (Mouse) 12371, (Rat) 58918
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.