Invitrogen
Manufacturer Code:PA578923
Catalog # PIPA578923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of rat Caspase-6 . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptotic protease Mch-2; CASP-6; Caspase-6; Caspase-6 CASP-6 Apoptotic protease Mch-2 Caspase-6 subunit p18 Caspase-6 subunit p11 Casp6 Mch2; Caspase-6 subunit p11; Caspase-6 subunit p18
Gene Aliases: CASP-6; Casp6; Mch2
UniProt ID: (Mouse) O08738
Entrez Gene ID: (Mouse) 12368, (Rat) 83584
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.