Novus Biologicals
Manufacturer Code:NBP270339AF405
Catalog # N270339A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 81-270 of human Caspase-4 (NP_150649) produced in E.coli. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: apoptotic cysteine protease Mih1/TX; apoptotic cysteine protease Mih1/TX CASP-4 caspase 4 apoptosis-related cysteine peptidase caspase 4 apoptosis-related cysteine protease caspase-4 EC 3.4.22 EC 3.4.22.57 ICE(rel)II ICE(rel)-II ICEREL-II ICH2 ICH-2 Mih1/TX Protease ICH-2 Protease TX TX; CASP-4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; Caspase-4; Caspase-4 subunit p10; Caspase-4 subunit p20; ICE and Ced-3 homolog 2; ICE(rel)-II; ICH-2; Mih1; protease ICH-2; Protease TX
Gene Aliases: CASP4; ICE(rel)II; ICEREL-II; ICH-2; ICH2; Mih1; Mih1/TX; TX
UniProt ID: (Human) P49662
Entrez Gene ID: (Human) 837
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.