Novus Biologicals
Manufacturer Code:NBP176812
Catalog # NBP176812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 15 amino acid peptide near the amino terminus of bovine Caspase-13. The immunogen is located within amino acids 80 - 130 of Caspase-13. Amino Acid Squence: ETVAGPDESVGSAAT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoptosis-related cysteine protease; apoptotic cysteine protease Mih1/TX CASP-4 caspase 4 apoptosis-related cysteine peptidase caspase 4 apoptosis-related cysteine protease caspase-4 EC 3.4.22 EC 3.4.22.57 ICE(rel)II ICE(rel)-II ICEREL-II ICH2 ICH-2 Mih1/TX Protease ICH-2 Protease TX TX; CASP-13; CASP-4; caspase 4, apoptosis-related cysteine protease; Caspase-13; Caspase-13 subunit 1; Caspase-13 subunit 2; Caspase-4; Caspase-4 subunit p10; Caspase-4 subunit p20; ERICE; Evolutionary related interleukin-1-beta-converting enzyme
Gene Aliases: CASP13; CASP4
UniProt ID: (Bovine) O75601
Entrez Gene ID: (Bovine) 338039
Molecular Function: cysteine protease enzyme modulator hydrolase protease protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.