Novus Biologicals
Manufacturer Code:NBP176602
Catalog # NBP176602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid synthetic peptide from the amino-terminus of human Caspase-4. The immunogen is located within amino acids 20 - 70 of Caspase-4. Amino Acid Squence: KEEEKKKYYDAKTEDK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoptotic cysteine protease Mih1/TX; apoptotic cysteine protease Mih1/TX CASP-4 caspase 4 apoptosis-related cysteine peptidase caspase 4 apoptosis-related cysteine protease caspase-4 EC 3.4.22 EC 3.4.22.57 ICE(rel)II ICE(rel)-II ICEREL-II ICH2 ICH-2 Mih1/TX Protease ICH-2 Protease TX TX; CASP-4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; Caspase-4; Caspase-4 subunit p10; Caspase-4 subunit p20; ICE and Ced-3 homolog 2; ICE(rel)-II; ICH-2; Mih1; protease ICH-2; Protease TX
Gene Aliases: CASP4; ICE(rel)II; ICEREL-II; ICH-2; ICH2; Mih1; Mih1/TX; TX
UniProt ID: (Human) P49662
Entrez Gene ID: (Human) 837
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.