Invitrogen
Manufacturer Code:PA539873
Catalog # PIPA539873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from human Caspase 4/5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoptotic cysteine protease Mih1/TX; CASP-4; CASP-5; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; caspase 5, apoptosis-related cysteine peptidase; caspase 5, apoptosis-related cysteine protease; Caspase-4; Caspase-4 subunit p10; Caspase-4 subunit p20; Caspase-5; Caspase-5 subunit p10; Caspase-5 subunit p20; ICE and Ced-3 homolog 2; ICE(rel)-II; ICE(rel)-III; ICE(rel)II ICEREL-II ICH-2 Mih1/TX TX CASP-4 ICE(rel)-II apoptotic cysteine protease Mih1/TX caspase 4 apoptosis-related cysteine protease caspase-4 protease ICH-2 protease TX ICE(rel)III ICEREL-III ICH-3 CASP-5 ICE(rel)-III TY protease caspase 5 apoptosis-related cysteine protease caspase-5 protease ICH-3 protease TY; ICH-2; Mih1; protease ICH-2; Protease ICH-3; Protease TX; Protease TY; TY protease
Gene Aliases: CASP4; CASP5; ICE(rel)II; ICE(rel)III; ICEREL-II; ICEREL-III; ICH-2; ICH-3; ICH2; ICH3; Mih1; Mih1/TX; TX
UniProt ID: (Human) P49662, (Human) P51878
Entrez Gene ID: (Human) 837, (Human) 838
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.